missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Wastewater Ultra Nucleic Acid Isolation Kit

Catalog No. A52606
Encompass
Change view
Click to view available options
Quantity:
100 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A52606 100 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A52606 Supplier Applied Biosystems™ Supplier No. A52606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit enables efficient, high-throughput nucleic acid extraction from wastewater samples.

  • The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit enables efficient, high-throughput nucleic acid extraction from wastewater samples
  • The kit utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid compatible with a broad range of applications
  • Wastewater and sewage surveillance is a useful tool for monitoring the regional spread of viral and bacterial pathogens
  • Surveillance at different levels of operatio has been shown to be a reliable way to detect the SARS-CoV-2 virus before it becomes widespread in a population
  • Wastewater surveillance can be used to monitor other viral targets such as influenza and norovirus
  • However, wastewater and sewage samples are some of the most challenging when it comes to extraction of RNA and DNA

Features of the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit include:

  • Automation-ready protocols designed for KingFisher Flex, KingFisher APEX, and KingFisher Duo systems
  • Fast procedure allows for 24–96 samples to be processed in ∼45 minutes
  • Compatible with wastewater and sewage concentrated via ultracentrifugation, precipitation, filtration, or bead-based viral enrichment and non-concentrated samples
  • Includes methods for 200 μL to 500 mL starting volumes of wastewater
  • Elution volumes ranging from 50 to 100 μL

Specifications

Content And Storage • 80 mL Lysis Buffer; room temperature
• 50 mL Binding Solution; room temperature
• 200 mL Wash Buffer; room temperature
• 25 mL Elution Solution; room temperature
• 4 mL Proteinase K; room temperature
• 2 mL DNA/RNA Binding Beads; room temperature

Isolation Technology Magnetic Bead
Sample Type Sludge, Wastewater
Elution Volume 50 to 200 μL
Final Product Type RNA and DNA
For Use With (Equipment) KingFisher™ Duo Prime, KingFisher™ Flex, KingFisher™ Apex
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization
High-throughput Compatibility High-throughput Compatible
Quantity 100 preps
Shipping Condition Room Temperature
Test Time Up to 29 min. when using automated protocol
Starting Material Amount Sludge: ≤200 mg
Wastewater: ≤500 mL
Show More Show Less
Is the purified viral nucleic acid that is obtained using MagMAX Wastewater Ultra Nucleic Acid Isolation Kit completely inhibitor-free?

The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit allows elimination of the vast majority of diverse inhibitors of enzymatic reactions, even from the most diluted and challenging samples such as wastewater. In very rare instances, if some inhibitors still remain within the purified nucleic acid, the sample can be diluted 10-20X prior to downstream PCR or other applications.

Is concentration of wastewater required prior to nucleic acid isolation?

Starting volumes of ≤1 mL can be processed using the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit without concentration. Larger wastewater volumes must first be concentrated using ultracentrifugation, precipitation, or concentration with Dynabeads (use the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit, with Virus Enrichment, Cat. No. A52610 for this protocol).

What starting volume of wastewater can be used with the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit?

The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit is compatible with starting volumes of 200 µL to 1 mL.

How long is the protocol for the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit?

The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit protocol takes 45 mins to get from sample to extracted nucleic acid.

Is the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit automation-compatible?

The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit is compatible with the KingFisher Flex, APEX, and Duo instruments, and can also be used for manual processing in 24- and 96-well plates.

For research use only. Not for use in diagnostic procedures.