missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen Binding Solution

Catalog No. A42359
Encompass
Change view
Click to view available options
Quantity:
550 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42359 550 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42359 Supplier Applied Biosystems™ Supplier No. A42359

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This binding buffer is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.

This binding buffer is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more binding buffer than is provided in the kit.

Specifications

Content And Storage Store at room temperature.

Sample Type Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Virus, Yeast
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX
Product Type Viral/Pathogen Binding Solution
Quantity 550 mL
Shipping Condition Room Temperature
Target Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids

For Research Use Only. Not for use in diagnostic procedures.