missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Saliva gDNA Wash I Solution

Catalog No. A39062
Encompass
Change view
Click to view available options
Quantity:
1.1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39062 1.1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39062 Supplier Applied Biosystems™ Supplier No. A39062
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

MagMAX Saliva gDNA Wash I Solution

This Wash I solution is a component of the MagMAX Saliva gDNA Isolation Kit.

This Wash I solution is a component of the MagMAX Saliva gDNA Isolation Kit. It is offered here as a stand-alone reagent for applications that require more Wash I solution than is provided in the kit.

Specifications

For Use With (Application) Next-Generation Sequencing, PCR
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX
Product Type Saliva gDNA Wash I Solution
Quantity 1.1 L
Shipping Condition Room Temperature
Target Genomic DNA
The MagMAX Saliva gDNA Lysis/Binding Solution and Wash I Solution have precipitates in them. Can I still use the buffers?

At lower temperatures, the components of the Lysis/Binding Solution and Wash I Solution may precipitate. To get rid of the precipitate, just warm the solutions to room temperature and invert bottles gently to solubilize the reagents before use.

Can I process less than 200 µL of saliva with the MagMAX Saliva gDNA Isolation Kit?

Yes you can, but the gDNA yields will be very low.

Will the MagMAX Saliva gDNA Isolation Kit work for sample types besides saliva?

No, this kit is specialized for freshly collected saliva and stabilized saliva samples only.

Do you sell the buffers and beads included in the MagMAX Saliva gDNA Isolation Kit as standalone items?

Yes, we offer the following kit components as stand-alone items to accommodate custom workflows or reagent volumes that have been optimized for your specific automation needs:
MagMAX Saliva gDNA Binding Beads: Cat. No. A39061
MagMAX Saliva gDNA Wash I Solution: Cat. No. A39062
MagMAX Saliva gDNA Lysis/Binding Solution: Cat. No. A39063
MagMAX Saliva gDNA Elution Solution: Cat. No. A39064

With the MagMAX Saliva gDNA Isolation Kit, do I use different protocols for processing fresh and stabilized saliva?

No, the same protocols are used for fresh and stabilized saliva.

Do you offer the Wash II Solution from the MagMAX Saliva gDNA Isolation Kit as a standalone item?

Wash II Solution is 80% ethanol and we do not offer it as a stand-alone item. It must be purchased separately from a different vendor.

How much saliva sample can I use with the MagMAX Saliva gDNA Isolation Kit?

You can use 50-500 µL of saliva in a 96-well plate. We also have protocols for processing 1 mL and 2 mL of saliva in a 24-well plate.


For Research Use Only. Not for use in diagnostic procedures.