Learn More
Applied BioSystems™ MagMAX™ Saliva gDNA Lysis/Binding Solution
Description
Includes
MagMAX Saliva gDNA Lysis/Binding Solution
This Lysis/Binding solution is a component of the MagMAX Saliva gDNA Isolation Kit. It is offered here as a stand-alone reagent for applications that require more Lysis/Binding solution than is provided in the kit.
Specifications
Specifications
| For Use With (Application) | Next-Generation Sequencing, PCR |
| High-throughput Compatibility | High-throughput Compatible, Manual Protocols, Automated Protocols |
| Product Line | MagMAX |
| Product Type | Saliva gDNA Lysis/Binding Solution |
| Quantity | 550 mL |
| Shipping Condition | Room Temperature |
| Target | Genomic DNA |
Frequently Asked Questions (FAQs)
At lower temperatures, the components of the Lysis/Binding Solution and Wash I Solution may precipitate. To get rid of the precipitate, just warm the solutions to room temperature and invert bottles gently to solubilize the reagents before use.
Yes you can, but the gDNA yields will be very low.
No, this kit is specialized for freshly collected saliva and stabilized saliva samples only.
Yes, we offer the following kit components as stand-alone items to accommodate custom workflows or reagent volumes that have been optimized for your specific automation needs:
MagMAX Saliva gDNA Binding Beads: Cat. No. A39061
MagMAX Saliva gDNA Wash I Solution: Cat. No. A39062
MagMAX Saliva gDNA Lysis/Binding Solution: Cat. No. A39063
MagMAX Saliva gDNA Elution Solution: Cat. No. A39064
No, the same protocols are used for fresh and stabilized saliva.
For Research Use Only. Not for use in diagnostic procedures.