Learn More
Applied Biosystems™ MagMAX™ Saliva gDNA Elution Solution
Description
Includes
MagMAX Saliva gDNA Elution Solution
Specifications
Specifications
| For Use With (Application) | Next-Generation Sequencing, PCR |
| High-throughput Compatibility | High-throughput Compatible, Manual Protocols, Automated Protocols |
| Product Line | MagMAX |
| Product Type | Saliva gDNA Elution Solution |
| Quantity | 120 mL |
| Shipping Condition | Room Temperature |
| Target | Genomic DNA |
Frequently Asked Questions (FAQs)
Yes you can, but the gDNA yields will be very low.
No, this kit is specialized for freshly collected saliva and stabilized saliva samples only.
Yes, we offer the following kit components as stand-alone items to accommodate custom workflows or reagent volumes that have been optimized for your specific automation needs:
MagMAX Saliva gDNA Binding Beads: Cat. No. A39061
MagMAX Saliva gDNA Wash I Solution: Cat. No. A39062
MagMAX Saliva gDNA Lysis/Binding Solution: Cat. No. A39063
MagMAX Saliva gDNA Elution Solution: Cat. No. A39064
Unfortunately, we cannot give out the exact formulation, but it is similar to low TE (10 mM Tris base, 0.1 mM EDTA) and is compatible with microarray, PCR, and NGS downstream applications.
No, the same protocols are used for fresh and stabilized saliva.
For Research Use Only. Not for use in diagnostic procedures.