Learn More
Applied Biosystems™ MagMAX™ Pro HT NoSpin Plasmid MiniPrep Kit
The MagMAX Pro HT NoSpin Plasmid Miniprep Kit enables scalable, ultra-rapid purification of high-quality, transfection-grade plasmid DNA (pDNA) from bacterial cultures. Using magnetic-bead technology, it simplifies workflows by eliminating centrifugation and vacuum filtration bottlenecks.
Supplier: Applied Biosystems™ A58311
Description
The Applied Biosystems MagMAX Pro HT NoSpin Plasmid Miniprep Kit provides a scalable and ultra-rapid approach for purifying high quality, transfection-grade plasmid DNA (pDNA) directly from bacterial culture. Utilizing advanced magnetic-bead technology, the MagMAX Pro HT NoSpin kit drastically simplifies and streamlines plasmid purification workflows by eliminating centrifugation and vacuum filtration bottlenecks.
MagMAX Pro HT NoSpin Plasmid Miniprep Kit features include:
- Bead-based—utilizes dual para-magnetic beads for plasmid isolation without time-intensive processing steps (centrifugation, vacuum filtration)
- Automation enabled—turn-key, automation-friendly protocols and scripts, allowing 96 samples to be processed in under 35 minutes on KingFisher Apex and KingFisher Flex systems
- Fast plasmid DNA isolation—35 minutes from start to completion and ready for downstream applications
- Wide range of compatibility—typical E. coli cultures, common medias, and range of plasmids all acceptable
- High-quality pDNA yields of up to 20 μg, ready for downstream applications
- Ready for transfection—isolated pDNA with very low endotoxin levels < 1 EU/μg
Specifically designed for high-throughput plasmid isolation, the MagMAX Pro HT NoSpin technology is automation friendly and suited for use with KingFisher Apex, KingFisher Flex, and KingFisher Presto purification systems, as well as integrated liquid handling robots.
The MagMAX Pro HT purification chemistry is both effective and ecofriendly. Kits include a novel endotoxin reduction buffer to remove pyrogenic contaminates, and buffers are free of ethanol and chaotropic salts.
pDNA purified with the MagMAX Pro HT NoSpin kit is suitable for a wide array of downstream applications, such as transfection, cloning, enzymatic restriction, in vitro transcription, and sequencing.
Chemical Identifiers
| 2°C to 8°C BactBind Beads, PlasmidPure Beads, Endotoxin Reduction Buffer 15°C to 25°C RNAse A, Resuspension Buffer, Lysis Buffer, Precipitation Buffer, Wash 1 Buffer, Wash 2 Buffer, Elution Buffer |
|
| Magnetic Bead | |
| Bacteria, E. coli | |
| KingFisher Apex, Flex and Presto Processors with 96 Deep Well Heads, Liquid Handling Robotic Systems | |
| Plasmid DNA (pDNA) | |
| Automated, pDNA extraction kit | |
| Transfection Grade | |
| ~ 32 min. for 96 samples, 1 KingFisher 96 DWP | |
| 5-12kb |
| 96 Deep-well Plate | |
| Mini | |
| 100 μL | |
| Cloning, Transfection, Protein Expression, Protein Production, mRNA production via In Vitro Transcription, Gene Editing, Sequencing, PCR, Nucleic acid labeling, PCR | |
| KingFisher 96 Deep Well Plates with or without barcodes | |
| 1 Ea. | |
| Bacterial Culture, 1 mL per well | |
| Up to 20 μg pDNA using high copy plasmid |
Specifications
| 2°C to 8°C BactBind Beads, PlasmidPure Beads, Endotoxin Reduction Buffer 15°C to 25°C RNAse A, Resuspension Buffer, Lysis Buffer, Precipitation Buffer, Wash 1 Buffer, Wash 2 Buffer, Elution Buffer |
|
| Magnetic Bead | |
| Bacteria, E. coli | |
| KingFisher Apex, Flex and Presto Processors with 96 Deep Well Heads, Liquid Handling Robotic Systems | |
| 1152 Reactions | |
| KingFisher 96 Deep Well Plates with or without barcodes | |
| 1 Ea. | |
| Bacterial Culture, 1 mL per well | |
| Up to 20 μg pDNA using high copy plasmid |
| 96 Deep-well Plate | |
| Mini | |
| 100 μL | |
| Cloning, Transfection, Protein Expression, Protein Production, mRNA production via In Vitro Transcription, Gene Editing, Sequencing, PCR, Nucleic acid labeling, PCR | |
| Plasmid DNA (pDNA) | |
| Automated, pDNA extraction kit | |
| Transfection Grade | |
| ~ 32 min. for 96 samples, 1 KingFisher 96 DWP | |
| 5-12kb |