missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Prime Viral/Pathogen Proteinase K

Catalog No. A58151
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A58151 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A58151 Supplier Applied Biosystems™ Supplier No. A58151

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MagMAX Prime Viral/Pathogen Proteinase K is offered separately here for applications that require more Proteinase K than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.

MagMAX Prime Viral/Pathogen Proteinase K is offered separately here for applications that require more Proteinase K than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.

Specifications

Bead Type Magnetic
Color Clear
Content And Storage Store at 15–25°C.
Format Bottle
Isolation Technology Beads
Sample Type Swabs in Transport Media, Saliva, Urine, Serum, Plasma
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Automated Protocols
Key Functions Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of Reactions Up to 2000 Reactions
Nucleic Acid Type DNA/RNA
Product Line MagMAX
Product Type Reagent
Quantity 10 mL
Sample Volume 200–400 μL
Shipping Condition Room Temperature
Target Viral, Bacterial, Parasitic, and Fungal Nucleic Acid
Show More Show Less