missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MagMAX™ Prime Viral/Pathogen Proteinase K Dye
Description
MagMAX Prime Viral/Pathogen Proteinase K Dye is offered separately here for applications that require more Proteinase K Dye than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.
Specifications
Specifications
| Bead Type | Magnetic |
| Color | Clear |
| Content And Storage | Store at 15–25°C. |
| Format | Tube |
| Isolation Technology | Beads |
| Sample Type | Swabs in Transport Media, Saliva, Urine, Serum, Plasma |
| For Use With (Application) | Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR |
| High-throughput Compatibility | High-throughput Compatible, Automated Protocols |
| Key Functions | Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid |
| No. of Reactions | Up to 2000 Reactions |
| Show More |