missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MagMAX™ Prime Stool Lysis Buffer
Description
MagMAX Prime Stool Lysis Buffer is specifically designed to enable efficient, high-throughput nucleic acid extraction from stool samples. It is designed to be used in conjunction with the MagMAX Prime bead-beating tubes and MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.
Specifications
Specifications
| Bead Type | Magnetic |
| Color | Clear |
| Content And Storage | Store at 15–25°C. |
| Format | Bottle |
| Isolation Technology | Beads |
| Sample Type | Fecal Swab in Transport Media, Stool |
| For Use With (Application) | Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR |
| High-throughput Compatibility | High-throughput Compatible, Automated Protocols |
| Key Functions | Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid |
| No. of Reactions | Up to 100 Reactions |
| Show More |
For Research Use Only.