missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosytstems™ MagMAX™ Prime G+ Bacteria and Fungal Lysis Buffer

Catalog No. A59053
Change view
Click to view available options
No. of Reactions:
Up to 100 Reactions
Up to 1000 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A59053 Up to 100 Reactions
A58153 Up to 1000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A59053 Supplier Applied Biosytstems™ Supplier No. A59053
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi.

MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi. It is designed to be used in conjunction with MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.

Specifications

Bead Type Magnetic
Color Clear
Content And Storage Store at –15 to –25°C.
Format Bottle
Isolation Technology Beads
Sample Type Swabs in Transport Media, Saliva, Urine, Serum, Plasma
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Automated Protocols
Key Functions Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of Reactions Up to 100 Reactions
Nucleic Acid Type DNA/RNA
Product Line MagMAX
Product Type Reagent
Quantity 5 mL
Sample Volume 200–400 μL
Shipping Condition Dry Ice
Target Viral, Bacterial, Parasitic, and Fungal Nucleic Acid
Show More Show Less