missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Prime Bead Beating Tubes

Catalog No. A58155
Change view
Click to view available options
Quantity:
100 tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A58155 100 tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A58155 Supplier Applied Biosystems™ Supplier No. A58155

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MagMAX Prime Bead Beating Tubes are specifically designed to enable efficient, high-throughput nucleic acid extraction from stool.

MagMAX Prime Bead Beating Tubes are specifically designed to enable efficient, high-throughput nucleic acid extraction from stool. They are designed to be used with MagMAX Prime Stool Lysis Buffer and MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.

Specifications

Bead Type Magnetic
Color Clear
Content And Storage Store at 15–25°C.
Format Tube
Isolation Technology Beads
Sample Type Fecal Swab in Transport Media, Stool
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Automated Protocols
Key Functions Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of Reactions 100 Reactions
Nucleic Acid Type DNA/RNA
Product Line MagMAX
Product Type Reagent
Quantity 100 tubes
Sample Volume 400 μL
Shipping Condition Room Temperature
Target Viral, Bacterial, Parasitic, and Fungal Nucleic Acid
Show More Show Less

For Research Use Only.