missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Microbiome Ultra Nucleic Acid Isolation Kit, with bead plate

Catalog No. a42357
Encompass
Change view
Click to view available options
Quantity:
100 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42357 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42357 Supplier Applied Biosystems™ Supplier No. A42357
Only null left

The MagMAX Microbiome Ultra Nucleic Acid Isolation Kit enables efficient, high-throughput nucleic acid extraction from stool and soil.

Stool and soil are often the sample types of choice for scientists in the microbiome field. However, these samples also happen to be some of the most challenging to extract RNA and DNA from, until now. The MagMAX Microbiome Ultra Nucleic Acid Isolation Kit enables efficient, high-throughput nucleic acid extraction from stool and soil. The kit utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid compatible with a broad range of applications, including microarrays, real-time PCR, and next-generation sequencing.

Features of the MagMAX Microbiome Nucleic Acid Isolation Kit include:
• Automation-ready protocols designed for the KingFisher Flex and KingFisher Duo Prime systems
• Fast procedure allows for 96 samples to be processed in ∼60 minutes
• Compatible with stool, soil, and swabs contained in transport medium
• Mechanical disruption via bead beating in 96-well plate or individual tubes
• Elution volumes ranging from 50 to 200 μL

Maximize sample input and nucleic acid recovery in minimal time
Up to 100 mg of stool can be processed per well in a KingFisher 96-deep well plate. When the extraction process is performed using a KingFisher Flex system, 96 samples can processed in ∼60 minutes; when using a KingFisher Duo Prime system 12 samples can be processed in ∼50 minutes. When the protocol is completed, pure RNA and DNA is concentrated in 100 μL of elution buffer.

Two bead beating formats available
High-throughput processing of difficult samples such as stool is possible with 96-deep well plates containing zirconia beads combined with optimized MagMAX Microbiome reagents. If processing 96 samples at a time is not needed then the same beads are available in tube format for lower-throughput processing. Once the bead beating step is complete, the lysate can then be transferred into a KingFisher 96-deep well plate and processed on a KingFisher Flex or KingFisher Duo Prime system.

Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the DNA more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with stool samples that are commonly used for microbiome research.

The Dynabeads MyOne Silane contained in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit have an optimized silica-like surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads MyOne Silane ideal for the recovery RNA and DNA. Once captured, these nucleic acids can then be eluted in of 50-100 μL of elution buffer for use in downstream applications.

Specifications

Content And Storage • 80 mL Lysis Buffer; room temperature
• 50 mL Binding Solution; room temperature
• 200 mL Wash Buffer; room temperature
• 20 mL Elution Solution; room temperature
• 4 mL Proteinase K; room temperature
• 2 mL Total Nucleic Acid Binding Beads; room temperature
• 1 96-Deep Well Plate; room temperature

Isolation Technology Magnetic Bead
Sample Type Bacteria, Biofluids, Fecal, Soil, Swabs (Buccal or Skin), Yeast
Elution Volume 50 to 200 μL
Final Product Type RNA and DNA
For Use With (Equipment) KingFisher™ Duo Prime, KingFisher™ Flex, KingFisher™ Apex
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization
High-throughput Compatibility High-throughput Compatible
Quantity 100 Preps
Shipping Condition Room Temperature
Test Time Up to 35 min. when using automated protocol
Starting Material Amount Bacteria: ≤500 μL
Biofluids: ≤500 μL
Fecal: ≤100 mg (200 μL for samples in storage solution)
Soil: ≤250 mg
Yeast: ≤500 μL
Show More Show Less
What are the bead beating options available for the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit?

Here are the different bead-beating options that can be used:

- Omni Bead Ruptor 96: 30 Hz for 2 min
- Mini Bead Beater 96: 2 min
- Bead Bug: 30 sec is enough but for higher yields, use for 4 min at 4 m/s
- Plate Shaker (Vortex with plate adapter): 5 min at 2,000 rpm
- Vortex with 24 tube adapter: 10 min at 2,500 rom

What types of downstream analysis are compatible with the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit?

The following downstream analysis are compatible with the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit: PCR, qPCR, qRT-PCR, OpenArray analysis, microarray analysis, and next-generation sequencing.

What are the throughput options available for the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit?

The MagMAX Microbiome Ultra Nucleic Acid Isolation Kit is available in two throughput formats: with bead tubes for low throughput and with bead plates for high throughput (96 samples at a time).

How long is the workflow for the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit?

For fecal samples, the complete workflow takes 50 min and for all other sample types, it takes 70 min.

What are my options to obtain DNA only or RNA only using the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit?

For DNA only: Add 10 µL of RNase A (Cat. No. AM2270, AM2271) in each well of the Wash 1 and Wash 2 plates.

For RNA only: DNA-free DNA Removal Kit (Cat. No. AM1906) can be used with purified total nucleic acid.

Is the purified DNA/RNA from the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit completely inhibitor-free?

The MagMAX Microbiome Ultra Nucleic Acid Isolation Kit is based on a newly developed buffer system, allowing elimination of the vast majority of diverse inhibitors of enzymatic reactions, even from the most challenging samples such as stool and soil. In very rare instances, if some inhibitors still remain within the purified DNA/RNA, the sample can be diluted 10-20X prior to downstream PCR or other reactions.

Does the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit efficiently lyse gram-positive bacteria and challenging fungal species?

The kit utilizes a combination of chemical and mechanical (bead beating) lysis strategies, which enables efficient lysis and DNA/RNA recovery from even the most challenging microbial species.

Which sample types can be processed with the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42357, A42358)?

This kit was developed for fast and efficient isolation of inhibitor-free microbial DNA/RNA from human stool, swabs (fecal, skin, buccal), samples in viral transport media (VTM), and body fluids (urine, saliva). It has also been confirmed to perform well with stool samples from other mammals, reptiles, fish, bird, and other samples like soil, bacterial cultures, and yeast cultures.

Considering that stool and soil are the most challenging sample types in terms of homogenization and depletion of inhibitors, the kit should work well for most other sample types that researchers might be processing in the lab.


For Research Use Only. Not for use in diagnostic procedures.