missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Dx Viral/Pathogen NA Isolation Kit

Catalog No. A52736
Change view
Click to view available options
No. of Reactions:
Up to 2,000 preps
Up to 600 preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A52736 Up to 600 preps
A52735 Up to 2,000 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A52736 Supplier Applied Biosystems™ Supplier No. A52736

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from respiratory biological specimens for downstream qPCR applications.

The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is specifically designed to recover viral and bacterial nucleic acids from human respiratory biological specimens for qPCR downstream applications. The kit uses MagMAX magnetic-bead technology to enable reproducible recovery of high-quality nucleic acids.

Features of the MagMAX Dx Viral/Pathogen NA Isolation Kit include:

  • For in vitro diagnostic use. This product is US IVD compliant
  • For isolation and purification of viral and bacterial nucleic acids from a range of respiratory biological specimens such as nasopharyngeal swab, anterior nasal swab, sputum, and saliva
  • Single, flexible automation protocol that accommodates sample volume inputs of 200–400 μL
  • Fast, automated procedures that allow 96 samples to be processed on the KingFisher Apex Dx Purification Instrument with ∼30-minute instrument run time

Access the Instructions for Use

(You will be redirected to sign into the resource library.)

Access the MagMAX Dx protocol for KingFisher Apex Dx instrument: Dx_MVP_RSP_APX_V1.kfdx

(You will be redirected to sign into the resource library.)

More information about the KingFisher Apex Dx Purification Instrument

Please check with your country specific representative for more information on product registration.

Specifications

Content And Storage

• Binding Solution, 160 mL
• Wash Solution, 300 mL
• Binding Beads, 6 mL
• Proteinase K, 3 mL
• Elution Buffer, 36 mL

Shipped and stored at room temperature.

Format 96-well Plate
Isolation Technology Magnetic Bead
Sample Type Nasopharyngeal swab, Anterior nasal swab, Saliva, Sputum
For Use With (Equipment) KingFisher™ Apex Dx
For Use With (Application) Real-Time PCR
No. of Reactions Up to 600 preps
Nucleic Acid Type Viral and Bacterial
Product Line MagMAX
Quantity Up to 600 preps
Shipping Condition Room Temperature
Target Respiratory Pathogens
Show More Show Less

For In Vitro Diagnostic Use