missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Dx Viral/Pathogen NA Isolation Kit

The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from respiratory biological specimens for downstream qPCR applications.

Supplier:  Applied Biosystems™ A52735

 View more versions of this product

Catalog No. A52735


Only null left

Description

Description

The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from respiratory biological specimens for downstream qPCR applications.

Features of the MagMAX Dx Viral/Pathogen NA Isolation Kit include:
• Compatible with multiple downstream assays
• Single purification protocol for routinely tested pathogens and respiratory biological specimen without need for additional lysis
• Ability to purify various samples on single plate
• Sustainable product with biodegradable ingredients
• Designed to work with KingFisher Apex Dx instrument in under 40 mins

The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in the techniques of automated magnetic-bead purification and in vitro diagnostic procedures.

Specifications

Chemical Identifiers

96-well Plate
Nasopharyngeal swab, Anterior nasal swab, Saliva, Sputum
Real-Time PCR
MagMAX
Respiratory Pathogens
Magnetic Bead
KingFisher™ Apex Dx
Viral and Bacterial
Room Temperature

Specifications

• Binding Solution, 530 mL
• Wash Solution, 1000 mL
• Binding Beads, 20 mL
• Proteinase K, 10 mL
• Elution Buffer, 120 mL

Shipped and stored at room temperature.

Magnetic Bead
KingFisher™ Apex Dx
Up to 2,000 preps
MagMAX
Room Temperature
96-well Plate
Nasopharyngeal swab, Anterior nasal swab, Saliva, Sputum
Real-Time PCR
Viral and Bacterial
Up to 2,000 preps
Respiratory Pathogens
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 Months

SDS
Documents

Documents

Promotions

Promotions

For In Vitro Diagnostic Use