missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra Kit

Catalog No. A25597
Encompass
Change view
Click to view available options
No. of Reactions:
2500 Preps
500 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A25597 500 Preps
A25598 2500 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25597 Supplier Applied Biosystems™ Supplier No. A25597

The MagMAX™ DNA Multi-Sample Ultra Kit is optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue.

The MagMAX™ DNA Multi-Sample Ultra Kit is optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue. gDNA purified with the kit is ideal for qPCR applications on sensitive, scalable platforms such as the QuantStudio™ 12K Flex System with OpenArray™ Block (with AccuFill™ System). The magnetic bead–based purification format allows you to easily scale from processing 12 to 500 samples a day, making it the ideal choice for pharmacogenomics (PGx) studies.

Features of the MagMAX™ DNA Multi-Sample Ultra Kit include:
• Streamlined protocols for numerous non-invasive biological samples
• Optimized to provide DNA yields suitable for OpenArray™ and Ion AmpliSeq™ applications
• Packaged for high-throughput needs

One Kit for a Diverse Set of Sample Types
Why purchase multiple purification kits to isolate gDNA from different sample types? The MagMAX™ DNA Multi-Sample Ultra Kit eliminates the headache of ordering these extra kits because it has been optimized for use with a variety of biological samples. The kit is designed for high-throughput isolation of gDNA from 1 buccal swab, 50 to 350 μL of whole blood, up to 400 μL urine (without sample concentration), up to 150 μL saliva, 1-5 mL mouth rinse, blood spot cards, and mammalian tissue samples (up to 10 mg). The protocols and magnetic bead format make the kit fully amenable to automation.

*Please note that the Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processor with a 24 Deep-well Head is recommended for sample volumes greater than 50 μL.

Benefits of MagMAX™ Magnetic Bead–based Purification
Magnetic beads offer many benefits compared to other technologies for isolating gDNA. Beads bind gDNA more efficiently than glass fiber filters, resulting in higher and more consistent gDNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples.

Accessory Products
Proteinase K (Cat. No. A25561), DNA Binding Beads (Cat. No. A25562), Wash Solution 1 Concentrate (Cat. No. AM8504), and Wash Solution 2 Concentrate (Cat. No. AM8640) are components of the kit that may also be purchased separately.

Specifications

Content And Storage • DNA Binding Beads, 1 × 8 mL (store at 2–8°C, DO NOT FREEZE)
• DNA Elution Buffer 1, 1 x 25 mL (store at room temperature)
• DNA Elution Buffer 2, 1 x 25 mL (store at room temperature)
• Wash Solution 1 Concentrate, 1 x 80 mL* (store at room temperature)
• Wash Solution 2 Concentrate, 1 x 162 mL* (store at room temperature)
• Multi-Sample DNA Lysis Buffer, 1 x 100 mL (store at room temperature)
• Nuclease-Free Water, 1 x 100 mL (store at room temperature)
• PK Buffer, 1 x 96 mL (store at room temperature)
• Proteinase K, 1 x 4 mL (store at -5 to -30°C)
• Ribonuclease A (RNase A), 1 x 1.25 mL (store at -5 to -30°C)
*after addition of alcohol
Format Kit
Isolation Technology Magnetic Bead
Sample Type PCR Products
For Use With (Application) Real-Time PCR, Sequencing
No. of Reactions 500 Preps
Product Line MagMAX
Product Type Kit, 500 preps
Quantity 500 Preps
What kind of samples can I use with the MagMAX DNA Multi-Sample Ultra Kit?

The MagMAX DNA Multi-Sample Ultra Kit is optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue.

Is there a specific protocol for preparing the lysate to be used with TaqPath ProAmp Mastermix (Cat. No. A30865)?

The standard protocols described in the user guides of the DNA Extract All Reagent Kit and MagMAX DNA Multi-Sample Ultra Kit should be followed.

What is the difference between Proteinase K for MagMAX DNA Multi-Sample Ultra Kit (Cat. No A25561) and Proteinase K for MagMAX 96 DNA Multi-Sample Kit (Cat. No. 4489110)?

The difference between Proteinase K for MagMAX‐96 DNA Multi‐Sample Kit and Proteinase K for MagMAX DNA Multi‐Sample Ultra Kit is the brand. The former is from Invitrogen legacy and the latter is from Applied Biosystems legacy. Both products have an identical concentration of 100 mg/mL and can be used interchangeably.

Where can I find a script for processing a large volume of blood using the MagMAX Multi-Sample Ultra kit (Cat. No. A25597) with the KingFisher Flex or KingFisher Duo Prime instruments?

Scripts for processing large volumes of whole blood (up to 1 mL) can be found on the MagMAX Multi-Sample Ultra kit product page under the Product Literature section at the bottom of the page. There are two scripts available:
• KingFisher DUO file: A25597_Large_Volume_Blood.bdz
• KingFisher Flex file: A25597_Large_Volume_Blood.bdz

*Use the page scrolling function to see the complete list of product literature for this product.

Do you have any positive controls for the vaginal microbiota assays?

The TaqMan Vaginal Microbiota Amplification Control (Cat. No. A32040) is a linearized multi-target plasmid with target sequences for each available vaginal microbiota profiling assay.

Which sample prep kit do you recommend when running urinary tract microbiota profiling experiments?

We recommend the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) for isolating DNA for urinary tract microbiota profiling experiments.

Which sample prep kit do you recommend when running vaginal microbiota profiling experiments?

We recommend the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) for isolating DNA for vaginal microbiota profiling experiments.

Do you have any positive controls for the urinary tract microbiota assays?

The TaqMan Urinary Tract Microbiota Amplification Control (Cat. No. A39174) is a linearized multi-target plasmid with target sequences for each available urinary tract microbiota profiling assay.

What type of samples is the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) optimized for?

We have optimized protocols for extracting DNA from buccal swabs, whole blood, blood cards (Whatman FTA), oral rinse, saliva, and urine samples. In addition, the kit can be used to process vaginal microbiota samples (e.g., ThinPrep, Surepath) for gDNA extraction from yeast, bacteria (Gram positive/negative), viruses, and protozoa with the addition of additional enzymatic steps.

In the MagMAX DNA Multi-Sample Ultra Kit protocol, why do blood samples appear cloudy after adding the Multi-Sample DNA Lysis Buffer?

On rare occasions, the sample may turn cloudy after adding the Multi-Sample DNA Lysis Buffer. The sample will clear up when isopropanol is added. This will not have any impact on the yield or quality of the DNA.

Can heparin be used to collect blood samples for the MagMAX DNA Multi-Sample Ultra Kit?

Heparin is not recommended as an anti-coagulant since it can cause inhibition of PCR reactions. Instead, we strongly recommend collecting blood samples in EDTA or sodium citrate.

What is the best way to remove the buccal swabs from the MagMAX DNA Multi-Sample Ultra Kit lysate?

We recommend transferring the lysate to a new MagMAX Express-96 Deep Well Plate (Cat. No. 4388476). This option eliminates contamination risks and saves time. To transfer lysates, set a multi-channel micropipetor to ˜300 µL and transfer one row at a time. Each well should contain 200-250 µL after transfer.

Can buccal swabs that are to be used with the MagMAX DNA Multi-Sample Ultra Kit be shipped after sample collection?

Buccal swabs can be shipped at 25 degrees C or below. Exposure of the swabs to high temperatures (over 30 degrees C) during shipment has been associated with poor performance in the workflow. To prevent degradation, ensure that buccal swabs are completely dry before shipment. Place the swabs in a paper envelope for storage after collection - paper envelopes are permeable and allow the swabs to dry effectively.

What buccal swabs do you recommended using with the MagMAX DNA Multi-Sample Ultra Kit?

We recommend using one of the following types of buccal swabs with foam tips:

- Puritan PurFlock Ultra Flocked Swabs (Fisher Scientific, Cat. No. 22-025-192)
- Puritan HydraFlock Swabs, standard tip (Puritan, Cat. No. 25-3306-H )
- Sterile Foam Tipped Swabs (Puritan, Cat. No. 25-1506 1PF)
- 4N6FLOQSwabs, regular tip (Thermo Fisher Scientific, Cat. No. 4473979)

Note: Please do not use cotton swabs, as they may contain PCR inhibitors.

Why does the MagMAX DNA Multi-Sample Ultra Kit have two DNA elution buffers?

DNA Elution Buffer 1 is at a basic pH to help maximize release of the gDNA from the magnetic beads and has to be neutralized by DNA Elution Buffer 2.

Why is no RNase treatment included in the MagMAX DNA Multi-Sample Ultra Kit workflow?

RNase A treatment is not critical for this workflow. RNA that might be present in the samples is likely to be degraded in the course of the experiment and should not interfere with downstream applications. For those sample types that might potentially have some RNA carryover, we have included the RNase A as part of the Bead mix as a precaution.

Can I combine DNA Elution Buffer 1 and DNA Elution Buffer 2 from the MagMAX DNA Multi-Sample Ultra Kit and just have a single elution step?

Yes, you can combine the two elution buffers prior to eluting the DNA; instead of having the instrument pause, requiring you to add DNA Elution Buffer 2, press ‘start' on the instrument - just be sure to combine them directly before adding to the elution plate and before putting the plate on the KingFisher instrument (you do not want them to sit as a mix for too long). For example, if you wish to elute in 50 µL of buffer, you can mix 25 µL of DNA Elution Buffer 1 and 25 µL of DNA Elution Buffer 2, then add the total 50 µL into a well and run the protocol as you normally would.

Can I edit the manufactured KingFisher Flex protocols?

No, manufactured protocols are locked for editing, but they can be renamed using the ‘save as' function and edited as a new version.

Can I use magnetic beads from other vendors with the KingFisher Flex instrument?

For nucleic acid extraction, we recommend using our MagMAX kits and MagJET kits. Other magnetic beads should work well as long as they are 0.5 to 10 µm in size. However, you will have to write your own KingFisher protocols using the Thermo Scientific BindIt software and validate/optimize the protocol on your own.

I am working with liquid samples with high volume and want to extract nucleic acid from them using the KingFisher Flex instrument. Can I use more volume than the protocol advises?

Yes, this can be done by sample splitting. Set the protocol so that the magnet will collect samples from multiple wells before continuing to the next step. Be sure not to overload.

Can I scale up my reaction volume with the KingFisher Flex instrument?

Yes, reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.

Are all of the Applied Biosystems MagMAX kits validated for the KingFisher Flex Magnetic Particle Processor?

Yes, all MagMAX kits can be used with the KingFisher Flex Magnetic Particle Processor.


For Research Use Only. Not for use in diagnostic procedures.