missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra 2.0 Binding Beads

Catalog No. A36579
Change view
Click to view available options
Quantity:
45 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A36579 45 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A36579 Supplier Applied Biosystems™ Supplier No. A36579
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

These Magnetic Binding beads are included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit.

These Magnetic Binding beads are included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit. They are available separately for applications that require more binding beads than are provided in the kit.

Specifications

Content And Storage Store at room temperature.
Product Line MagMAX
Product Type DNA Multi-Sample Ultra 2.0 Binding Bead
Quantity 45 mL
Shipping Condition Room Temperature
Are there larger volume standalone reagent components available to scale up the MagMAX HMW DNA workflow?

Yes, the following larger volume standalone components are available for purchase separately:
- MagMAX HMW DNA Booster, 1,000 preps (1.0 mL) (Cat. No. A36625)
- MagMAX DNA Multi-Sample Ultra 2.0 Proteinase K (45 mL) (Cat. No. A36578)
- MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (45 mL) (Cat. No. A36579)
- MagMAX DNA Multi-Sample Ultra 2.0 Wash I Solution (1.1 L) (Cat. No. A36580)
- MagMAX DNA Multi-Sample Ultra 2.0 Lysis Binding Solution (450 mL) (Cat No. A36581)
- MagMAX DNA Multi-Sample Ultra 2.0 Elution Solution (120 mL) (Cat No. A36582)
- MagMAX DNA Multi-Sample Ultra 2.0 Enhancer Solution (45 mL) (Cat No. A36583)
- MagMAX Cell and Tissue DNA Extraction Buffer (600 mL) (Cat No. A45470)

If I have already purchased the MagMAX DNA Multi-Sample Ultra 2.0 Kit or standalone components, what do I need to extract high molecular weight DNA?

In addition to the MagMAX DNA Mult-Sample Ultra 2.0 product(s) you will need:
MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624) or
In addition to the MagMAX DNA Mult-Sample Ultra 2.0 product(s) you will need:
MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624) or MagMAX HMW DNA Booster, 1,000 preps (Cat No. A36625).

For cell and tissue samples include:
MagMAX Cell and Tissue DNA Extraction Buffer, 100 preps (Cat. No. A45469) or MagMAX Cell and Tissue DNA Extraction Buffer, 1,000 preps (Cat. No. A45470)
The MagMAX HMW DNA User Guide (MAN1001478)
The specific HMW DNA KingFisher automation protocol for your KingFisher Purification System (see product description page for download).
MagMAX HMW DNA Booster, 1,000 preps (Cat No. A36625).

For cell and tissue samples include:
MagMAX Cell and Tissue DNA Extraction Buffer, 100 preps (Cat. No. A45469) or MagMAX Cell and Tissue DNA Extraction Buffer, 1,000 preps (Cat. No. A45470)
The MagMAX HMW DNA User Guide (MAN1001478)
The specific HMW DNA KingFisher automation protocol for your KingFisher Purification System (see product description page for download).

Can the MagMAX HMW DNA Booster be paired with other genomic DNA isolation kits?

The MagMAX HMW DNA Booster is specifically designed and optimized for use with the MagMAX DNA Multi-Sample Ultra 2.0 (Cat. No. A36570) chemistry.

Are the DNA Binding Beads included within the MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (Cat. No. A36570) available as a stand-alone product and in a smaller volume?

We offer the MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (Cat. No. A36579), but the volume is 45 mL. The volume of the beads in the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) is 4.5 mL.

For Research Use Only. Not for use in diagnostic procedures.