Learn More
Applied Biosystems™ MagMAX™ CORE Mechanical Lysis Module & Glass Microbeads Bundle with CORE Chemistry
Description
MagMAX™ CORE Glass Microbeads are a supplement to the MagMAX™ CORE Nucleic Acid Purification Kit and MagMAX™ CORE Mechanical Lysis Module. The beads aid in the homogenization of completely desiccated feces such as environmental samples or fecal pellets.
The Mechanical Lysis Module is also available separately.
Specifications
Specifications
| Content And Storage | • 1 x 50 mL MagMAX CORE Glass Microbeads • 1 x 50 mL MagMAX CORE Lysis Solution • 1 x 45 mL MagMAX CORE Binding Solution • 1 x 60 mL MagMAX CORE Wash Solution 1 • 1 x 60 mL MagMAX CORE Wash Solution 2 • 1 x 12 mL MagMAX CORE Elution Buffer • 1 x 2.2 mL MagMAX CORE Magnetic Beads • 1 x 1.25 mL MagMAX CORE Proteinase K • 1 x 45 mL MagMAX CORE Clarifying Solution • 100 MagMAX CORE Bead Beating Tubes (2 mL) |
| Sample Type | Bacteria, Blood, Buccal Samples, Cells, DNA, RNA, Tissue, Virus |
| For Use With (Application) | Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR |
| High-throughput Compatibility | Automated Protocols, Manual Protocols, Multiplexing |
| Product Line | MagMAX |
| Quantity | 100 Tests |
| Shipping Condition | Room Temperature |
| Target | Total RNA, Pathogen DNA/RNA, Genomic DNA, Viral DNA/RNA |
Frequently Asked Questions (FAQs)
The only difference is that the MagMAX CORE Mechanical Lysis Module & Glass Microbeads Bundle with CORE Chemistry (Cat. No. A37488) contains glass microbeads, whereas the other kit (Cat. No. A37487) does not. The beads are also available as a standalone item (Cat. No. A37489) and aid in the homogenization of completely desiccated fecal or environmental samples. These glass microbeads are added to the desiccated sample together with some deionized water, vortexed, and incubated for about 10 minutes prior to the bead beating step.
The size of the bead tubes that come with these two kits is different, but the bead beating settings are the same. The Mechanical Lysis Module contains a workflow for up to 5 g of feces in addition to the standard 0.3 g protocol.
The MagMAX CORE Mechanical Lysis Module is optimized for isolating nucleic acid from difficult to-lyse targets such as mycobacterium (specifically Mycobacterium avium paratuberculosis and tuberculosis), spores, and complex samples used for microbiome testing.
For Research Use Only