missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ CORE Glass Microbeads

Catalog No. A37489
Encompass
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A37489 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A37489 Supplier Applied Biosystems™ Supplier No. A37489

MagMAX™ CORE Glass Microbeads are offered as a supplement to the MagMAX™ CORE Nucleic Acid Purification Kit and the MagMAX™ CORE Mechanical Lysis Module.

MagMAX™ CORE Glass Microbeads aid in the homogenization of completely dessicated feces such as environmental samples or fecal pellets.

For a kit that contains these microbeads as well as the MagMAX™ CORE Nucleic Acid Purification Kit and the MagMAX™ CORE Mechanical Lysis Module, please see Cat. No. A37488.

Specifications

Content And Storage 50 mL bottle, store at room temperature
Sample Type Blood, Buccal Samples, Cells, DNA, RNA, Tissue, Virus
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility Manual Protocols, Multiplexing
Product Line MagMAX
Quantity 100 Tests
Shipping Condition Room Temperature
Target Total RNA, Pathogen DNA/RNA, Genomic DNA, Viral DNA/RNA

For Research Use Only