missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ CORE ™ Disruption Modules

Catalog No. A40004819
Change view
Click to view available options
Format:
Plate
Tube
Quantity:
100 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Quantity
A40004819 Tube 100 Reactions
A40004818 Plate 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A40004819 Supplier Applied Biosystems™ Supplier No. A40004819

The Applied Biosystems MagMAX CORE disruption modules are a sample preparation system designed to complement the MagMAX CORE Nucleic Acid Purification Kit.

The Applied Biosystems MagMAX CORE disruption modules are a sample preparation system designed to complement the MagMAX CORE Nucleic Acid Purification Kit. This modular technology is optimized for the purification of nucleic acids from a wide range of sample matrices commonly submitted for veterinary diagnostic testing. The MagMAX CORE disruption modules aid in the processing of hard-to-lyse organisms like Mycobacterium avium paratuberculosis (MAP).

Key features

  • Convenience—a complete solution using CORE chemistry
  • Confidence—optimized extraction of nucleic acids from difficult-to-lyse samples
  • Flexible—purchase only what you need to lyse MAP and other hard-to-lyse organisms; 100 reaction tube module for lower throughput or 500 reaction plate module for higher throughput
  • Ergonomy—streamlined sample preparation protocol versus the previous mechanical lysis module
  • Sustainability—reduced tube consumption with use of high-throughput plate module

There are two options of module depending on workflow and throughput needs:

  • MagMAX CORE Disruption Tubes Module, 100 reactions, contains bead beating tubes prefilled with zirconia beads for effective disruption of individual samples
  • MagMAX CORE Disruption Plates Module, 500 reactions, contains 96-deep-well plates prefilled with zirconia beads and foil seal adapted for effective disruption of a higher number of samples

Specifications

Detection Method qPCR
Format Tube
Isolation Technology Mechanical Disruption
Scale Maxi
For Use With (Application) DNA Purification
Quantity 100 Reactions

For Research Use Only.