missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Cell-Free DNA Lysis/Binding Buffer

Catalog No. A33600
Encompass
Change view
Click to view available options
Quantity:
425 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33600 425 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A33600 Supplier Applied Biosystems™ Supplier No. A33600
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

This buffer is included in the MagMAX Cell-Free DNA Isolation Kit.

This buffer is included in the MagMAX Cell-Free DNA Isolation Kit (Cat. No. A29319). It is made available separately for applications that require more Lysis/Binding Buffer than is provided in the kit.

Specifications

Sample Type Blood
For Use With (Application) Next-Generation Sequencing, Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX
Product Type DNA Lysis/Binding Buffer
Quantity 425 mL
Shipping Condition Room Temperature
Target Genomic DNA

For Research Use Only. Not for use in diagnostic procedures.