missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva MAbTrap™ Kit
Description
Includes
One HiTrap Protein G HP 1mL column with 50mL 10X binding buffer, 15mL 10X elution buffer, 25mL neutralizing buffer, connectors, and syringe
MAbTrap Kit contains everything necessary to perform a complete one step purification without using complicated equipment, eliminating time-consuming buffer preparation and column packing. The kit includes one HiTrap column, prepacked with Protein G Sepharose High Performance with sufficient buffer concentrate for 20 purifications.
- MAbTrap Kit contains everything necessary to perform a complete one-step purification without using complex equipment
- Convenient kit for purification of IgG from monoclonal and polyclonal material using a syringe in less than 15 min.
- Kit contains one HiTrap Protein G HP column prepacked with Protein G Sepharose High Performance and enough buffer concentrate for 20 preparations with a syringe
- Recombinant protein G lacking albumin binding region
Specifications
Specifications
| Bed Volume | 1 mL |
| Description | MAbTrap is an affinity chromatography kit, with everything required for small-scale IgG purification from ascites fluid, serum, and cell culture supernatants. |
| Flow Rate | 4 mL/min. |
| Content And Storage | 4 to 8°C, 20% Ethanol |
| Quantity | 1 Kit |
| For Use With (Application) | For purification of IgG from monoclonal and polyclonal material using a syringe in <15 min. |
| Type | MAbTrap Kit |
| Binding Capacity | >25 mg/mL Human IgG Medium |