missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MabSelect PrismA Chromatography Resin

Catalog No. 09928074
Change view
Click to view available options
Quantity:
25 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09928074 200 mL
09928073 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09928074 Supplier Cytiva Supplier No. 17549802

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protein A Chromatography Resin

MabSelect PrismA has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing future demands in mAb processing to be met.

Specifications

Matrix Highly cross-linked agarose, spherical.
Content And Storage 2 to 8°C, 20% ethanol
Max. Operating Pressure 300 cm/hr.
Particle Size 60 μm
Quantity 200 mL
For Use With (Application) Antibody affinity chromatography
Type Chromatography Resin
Ligand Type Alkaline-stabilized protein A-derived (E.coli).
Binding Capacity 80 mg Human IgG/mL Resin at 6 min residence time. 65mg Human IgG/mL Resin at 4 min residence time.