missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns
GREENER_CHOICE

Catalog No. DX075592
Change view
Click to view available options
Length (Metric):
50 mm
150 mm
300 mm
Diameter (Metric):
2.1 mm
4 mm
7.8 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
DX075592 150 mm 4 mm
13622467 300 mm 7.8 mm
DX074696 300 mm 4 mm
DX074697 50 mm 4 mm
03898145 150 mm 2.1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. DX075592 Supplier Thermo Scientific™ Supplier No. 075592

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use these SEC columns for monoclonal antibody (mAb) analysis, including the high-resolution separation of monomers, aggregates and fragments.

Use Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns for high-resolution separation of monoclonal antibody (mAb) analysis, including the monomers, aggregates and Fab and Fc fragments resulting from proteolysis. These size-exclusion chromatography (SEC) columns provide superior performance, even under nondenaturing conditions, and in high- and low-salt mobile phases and volatile eluents.

High-Performance SEC Columns for MAb Analysis

  • Hydrophilic bonded layer minimizes undesired interactions between the biomolecules and the stationary phase
  • Nonmetallic and biocompatible PEEK housing eliminates metal contamination from the column hardware
  • Stable surface bonding leads to low column bleed and compatibility with MS, ELSD and Thermo Scientific™ Dionex™ Corona™ CAD detection
  • Rugged column packing provides reliability, reproducibility and long column life
  • Columns are available in 5μm particle size and 150 and 300mm lengths, with an average pore diameter of 300Å

Specifications

Column Chemistry Proprietary Diol
Column Format Analytical Column
Diameter (Metric) 4 mm
Length (Metric) 150 mm
Max. Pressure 600 psi (40 bar)
Particle Size 5 μm
pH 2.5 to 7.5
Pore Size 300 Å
Quantity Each
Solvents 100% Organic Solvents
Temperature 30°C
USP Type L59
Stationary Phase Diol (OH)
Product Line MAbPac
Packing Material Spherical, Fully Porous, Ultrapure Silica
Column Type Size Exclusion
Phase Size Exclusion
Material PEEK
Technique Bio-LC
Green Features FSC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.