Learn More
Thermo Scientific™ MAbPac™ Reversed Phase HPLC Columns
Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.
Supplier: Thermo Scientific™ 088645
Description
Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.
Compatible with organic solvent used in HPLC and MS under RP conditions- Resin is based on a hydrophobic supermacroporous 4μm polymer particles
- Stable at extreme pH (0 to 14) and high temperature (up to 110 °C)
- 1,500Å large pore size polymeric resin
- Efficient separation of protein molecules with very low carry over
- Superior resolution power for monoclonal antibodies and related substances
- High efficiency with low carry-over
- Excellent MS compatibility
- Wide operating pH range: 0 to 14
- High temperature stability: up to 110°C
- High throughput
Specifications
| Analytical Column | |
| 50 mm | |
| 4 μm | |
| 1500 Å | |
| <110°C | |
| MAbPac | |
| Reversed Phase | |
| Bio-LC |
| 3 mm | |
| 4000 psi (275 bar) | |
| 0 to 14 | |
| Each | |
| Phenyl (Ph) | |
| DVB (Divinylbenzene) Polymer | |
| Reversed Phase | |
| Responsibly packaged |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.