missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ Protein A Antibody Analysis and Purification HPLC Column, 12 μm, 4 mm x 35 mm
GREENER_CHOICE

Catalog No. DX082539
Change view
Click to view available options
Length (Metric):
35 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
DX082539 35 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DX082539 Supplier Thermo Scientific™ Supplier No. 082539

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Column Format: Analytical Column; Pore Size: Nonporous; Column Phase: Affinity Separation

Run fast, accurate titer analysis of monoclonal antibodies (mAbs) in harvest cell cultures with the Thermo Scientific™ MAbPac™ Protein A LC Columns. This column provides high throughput and accurate analyses through a combination of low backpressure and high efficiency. Its unique format makes automation of loading, binding, elution and collection using biocompatible systems quick and easy. A nonporous, polymeric resin with a hydrophobic divinylbenzene core and a hydrophilic surface optimized for affinity separation makes this column rugged and reproducible.

Fast, Accurate mAb Titer Analysis

  • Complete cycle time, including equilibration, of two minutes
  • Linear over a wide concentration range (0.025 to 5mg/mL) without carryover
  • Low column pressure of less than 250 psi at 2mL/min
  • Excellent sample recovery

Use the MAbPac Protein A LC column for both titer analysis or concentrating and purifying IgG material for later experiments or two-dimensional (2D) chromatography. It is available in an HPLC bio-inert, 4×35mm PEEK column body.

Specifications

Column Format Analytical Column
Diameter (Metric) 4 mm
Length (Metric) 35 mm
Max. Pressure 1000 psi (70 bar)
Particle Shape Spherical polymer
Particle Size 12 μm
pH 2.5 to 7.5
Pore Size Nonporous
Quantity Each
Temperature 30°C
Stationary Phase Protein A
Product Line MAbPac
Packing Material Spherical, Nonporous Polymer
Column Type Antibody Analysis and Purification
Phase Affinity Separation
Technique Bio-LC
Green Features FSC
Product Type Analytical Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.