Learn More
Thermo Scientific™ MAbPac™ HIC-Butyl HPLC Columns
Separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs) with excellent biocompatibility, low carryover and high resolution.
$645.11 - $3788.13
Specifications
| Max. Pressure | 4000 psi (275 bar) |
|---|---|
| Particle Size | 5 μm |
| pH | 2 to 12 |
| Pore Size | Nonporous |
| Quantity | Each |
| Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
|
03-898-158
|
Thermo Scientific™
088559 |
10 mm | 4.6 mm |
Each for $645.11
|
|
||||
|
03-898-157
|
Thermo Scientific™
88460 |
100 mm | 4.6 mm |
Each for $3,788.13
|
|
||||
Description
Use the Thermo Scientific™ MAbPac™ HIC-Butyl HPLC columns to separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs). This column is based on hydrophilic, nonporous, 5μm polymer particles functionalized with butyl groups. The column's hydrophilic resin provides excellent biocompatibility and low carryover, while the optimal density of the butyl groups provides high resolution separation.
Features of MabPac HIC-Butyl Columns- Optimal selectivity for ADCs
- High resolution and high efficiency
- Stable at wide range of pH (2-12)
- Low carryover
- Rugged column packing
Specifications
| 4000 psi (275 bar) | |
| 2 to 12 | |
| Each | |
| MAbPac | |
| Hydrophobic Interaction | |
| Responsibly packaged |
| 5 μm | |
| Nonporous | |
| 60°C | |
| Hydrophilic Polymer | |
| Hydrophobic Interaction |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.