Learn More
Thermo Scientific™ MAbPac™ HIC-20 HPLC Columns

Description
Use this column to separate monoclonal antibodies (mAbs), mAb fragments and oxidized variants by hydrophobic interaction chromatography (HIC).
Use the Thermo Scientific™ MAbPac™ HIC-20 HPLC column for separation of monoclonal antibodies (mAbs), mAb fragments and oxidized variants by hydrophobic interaction chromatography (HIC). The unique chemistry of this silica column offers high resolution and excellent selectivity. The advanced surface bonding technology provides excellent chemical stability, high resolution, and unique selectivity optimized for mAbs and different from other HIC phases.
Characteristics of the MAbPac HIC-20 Column
This column is based on high-purity, spherical, wide-pore (1,000 Å), 5μm silica particles functionalized with proprietary alkyl amide groups offering:
- Optimal selectivity for mAbs and related variants
- Excellent separation of mAb fragments and oxidized variants
- High resolution and high efficiency
- Compatible with both organic solvent and aqueous mobile phase
- Rugged column stability
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 100 mm |
| Max. Pressure | 6000 psi (415 bar) |
| Particle Size | 5 μm |
| pH | 2 to 9 |
| Pore Size | 1000 Å |
| Quantity | Each |
| Temperature | 60°C |
| Stationary Phase | Silica |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.