missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva M13KO7 Helper Phage
Description
- The phage is an M13mpl derivative containing a modified gene 11
- Color: Colourless to light yellow
Specifications
Specifications
| Concentration | >5 x 1010 pfu/mL |
| Color | Colorless to light yellow |
| Quantity | 100 μL |
| Form | Liquid |