Learn More
m-Tolylhydrazine hydrochloride, 98%
CAS: 637-04-7 | C7H11ClN2 | 158.63 g/mol
Supplier: Thermo Scientific Chemicals A1366706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| m-Tolylhydrazine hydrochloride | |
| ∼193°C (decomposition) | |
| MFCD00012932 | |
| UN2811 | |
| 3-methylphenylhydrazine hydrochloride, m-tolylhydrazine hydrochloride, m-tolylhydrazinium 1+ chloride, 3-methylphenyl hydrazine hydrochloride, 3-methylphenylhydrazine hcl, m-tolyhydrazine hydrochloride, 1-3-methylphenyl hydrazine hydrochloride, 3-tolylhydrazine hydrochloride, 3-methyl phenylhydrazine hydrochloride, hydrazine, 3-methylphenyl-, monohydrochloride | |
| [H+].[Cl-].CC1=CC=CC(NN)=C1 | |
| 158.63 | |
| 158.63 |
| 637-04-7 | |
| C7H11ClN2 | |
| 5 g | |
| 3563995 | |
| SHTBTGXNVWYODU-UHFFFAOYSA-N | |
| (3-methylphenyl)hydrazine;hydrochloride | |
| 71562 | |
| 98% |
Chemical Identifiers
| 637-04-7 | |
| 158.63 | |
| SHTBTGXNVWYODU-UHFFFAOYSA-N | |
| 71562 | |
| [H+].[Cl-].CC1=CC=CC(NN)=C1 |
| C7H11ClN2 | |
| MFCD00012932 | |
| 3-methylphenylhydrazine hydrochloride, m-tolylhydrazine hydrochloride, m-tolylhydrazinium 1+ chloride, 3-methylphenyl hydrazine hydrochloride, 3-methylphenylhydrazine hcl, m-tolyhydrazine hydrochloride, 1-3-methylphenyl hydrazine hydrochloride, 3-tolylhydrazine hydrochloride, 3-methyl phenylhydrazine hydrochloride, hydrazine, 3-methylphenyl-, monohydrochloride | |
| (3-methylphenyl)hydrazine;hydrochloride |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 211-276-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only