missing translation for 'onlineSavingsMsg'
Learn More

m-Tolylhydrazine hydrochloride, 98%

Catalog No. p-7021362
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1366706 5 g
AAA1366714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1366706 Supplier Thermo Scientific Chemicals Supplier No. A1366706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 637-04-7 | C7H11ClN2 | 158.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 637-04-7
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.63
MDL Number MFCD00012932
InChI Key SHTBTGXNVWYODU-UHFFFAOYSA-N
Synonym 3-methylphenylhydrazine hydrochloride, m-tolylhydrazine hydrochloride, m-tolylhydrazinium 1+ chloride, 3-methylphenyl hydrazine hydrochloride, 3-methylphenylhydrazine hcl, m-tolyhydrazine hydrochloride, 1-3-methylphenyl hydrazine hydrochloride, 3-tolylhydrazine hydrochloride, 3-methyl phenylhydrazine hydrochloride, hydrazine, 3-methylphenyl-, monohydrochloride
PubChem CID 71562
IUPAC Name (3-methylphenyl)hydrazine;hydrochloride
SMILES [H+].[Cl-].CC1=CC=CC(NN)=C1

Specifications

Melting Point ∼193°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 3563995
Formula Weight 158.63
Percent Purity 98%
Chemical Name or Material m-Tolylhydrazine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 211-276-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only