missing translation for 'onlineSavingsMsg'
Learn More

m-Toluidine-4-sulfonic Acid 95.0+%, TCI America™

Catalog No. A032925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A032925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A032925G Supplier TCI America Supplier No. A032925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 133-78-8
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.21
MDL Number MFCD00035774
InChI Key ZDIRCGKEOWZBIM-UHFFFAOYSA-N
Synonym 5-Aminotoluene-2-sulfonic Acid, 3-Methylaniline-4-sulfonic Acid, 4-Amino-2-methylbenzenesulfonic Acid
PubChem CID 67248
IUPAC Name 4-amino-2-methylbenzene-1-sulfonic acid
SMILES CC1=CC(N)=CC=C1S(O)(=O)=O

Specifications

Quantity 25 g
Formula Weight 187.21
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material m-Toluidine-4-sulfonic Acid
EINECSNumber (3)-1978
RTECSNumber XT6325000
TSCA No