missing translation for 'onlineSavingsMsg'
Learn More

m-Toluic Acid 98.0+%, TCI America™

Catalog No. T029125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T029125G 25 g
T0291500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T029125G Supplier TCI America Supplier No. T029125G

Chemical Identifiers

CAS 99-04-7
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002523
InChI Key GPSDUZXPYCFOSQ-UHFFFAOYSA-N
Synonym m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext
PubChem CID 7418
ChEBI CHEBI:10589
IUPAC Name 3-methylbenzoic acid
SMILES CC1=CC=CC(=C1)C(O)=O

Specifications

Melting Point 112°C
Boiling Point 106°C
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material m-Toluic Acid
EINECSNumber (3)-1285
RTECSNumber XU1200000
TSCA Yes