Learn More
m-Toluic acid, 99%
CAS: 99-04-7 | C8H8O2 | 136.15 g/mol
Supplier: Thermo Scientific Chemicals A137850B
Description
m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsm-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.
Solubility
Soluble in alcohol and ether.
Notes
Incompatible with strong oxidizing agents. Store away from incompatible materials such as oxidizing agents.
Specifications
| m-Toluic acid | |
| 108°C to 110°C | |
| 263°C | |
| C8H8O2 | |
| 1000 g | |
| 14,9535 | |
| Soluble in alcohol and ether. | |
| CC1=CC=CC(=C1)C(O)=O | |
| 136.15 | |
| CHEBI:10589 | |
| 99% |
| 99-04-7 | |
| 1.05 | |
| 150°C (302°F) | |
| MFCD00002523 | |
| 970526 | |
| m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext | |
| GPSDUZXPYCFOSQ-UHFFFAOYSA-N | |
| 3-methylbenzoic acid | |
| 7418 | |
| 136.15 |
Chemical Identifiers
| 99-04-7 | |
| 136.15 | |
| GPSDUZXPYCFOSQ-UHFFFAOYSA-N | |
| 7418 | |
| 3-methylbenzoic acid |
| C8H8O2 | |
| MFCD00002523 | |
| m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext | |
| CHEBI:10589 | |
| CC1=CC=CC(=C1)C(O)=O |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P280-P305+P351+P338-P310
H318
EINECSNumber : 202-723-9
RTECSNumber : XU1200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only