missing translation for 'onlineSavingsMsg'
Learn More

m-Toluic acid, 99%

Catalog No. AAA1378530
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1378530 250 g
AAA137850B 1000 g
AAA137850I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1378530 Supplier Thermo Scientific Chemicals Supplier No. A1378530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 99-04-7 | C8H8O2 | 136.15 g/mol

m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.

Solubility
Soluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents. Store away from incompatible materials such as oxidizing agents.

Chemical Identifiers

CAS 99-04-7
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002523
InChI Key GPSDUZXPYCFOSQ-UHFFFAOYSA-N
Synonym m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext
PubChem CID 7418
ChEBI CHEBI:10589
IUPAC Name 3-methylbenzoic acid
SMILES CC1=CC=CC(=C1)C(O)=O

Specifications

Melting Point 108°C to 110°C
Density 1.05
Boiling Point 263°C
Flash Point 150°C (302°F)
Quantity 250 g
Beilstein 970526
Merck Index 14,9535
Solubility Information Soluble in alcohol and ether.
Formula Weight 136.15
Percent Purity 99%
Chemical Name or Material m-Toluic acid
Show More Show Less
Hazard Category H318
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 202-723-9
RTECSNumber XU1200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only