missing translation for 'onlineSavingsMsg'
Learn More

m-Toluenethiol 98.0+%, TCI America™

Supplier:  TCI America T028925G

 View more versions of this product

Catalog No. T028925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

m-Toluenethiol
Yellow
C7H8S
25 g
m-thiocresol, m-toluenethiol, 3-methylthiophenol, 3-thiocresol, benzenethiol, 3-methyl, m-methylbenzenethiol, m-methylthiophenol, 3-mercaptotoluene, m-tolylmercaptan, m-mercaptotoluene
CC1=CC=CC(S)=C1
124.20
124.20
Liquid
108-40-7
195°C
MFCD00004843
2810
WRXOZRLZDJAYDR-UHFFFAOYSA-N
3-methylbenzene-1-thiol
7930
≥98.0% (GC,T)

Chemical Identifiers

108-40-7
124.20
WRXOZRLZDJAYDR-UHFFFAOYSA-N
7930
CC1=CC=CC(S)=C1
C7H8S
MFCD00004843
m-thiocresol, m-toluenethiol, 3-methylthiophenol, 3-thiocresol, benzenethiol, 3-methyl, m-methylbenzenethiol, m-methylthiophenol, 3-mercaptotoluene, m-tolylmercaptan, m-mercaptotoluene
3-methylbenzene-1-thiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2895

RTECSNumber : XT8700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions