missing translation for 'onlineSavingsMsg'
Learn More

m-Toluenethiol 98.0+%, TCI America™

Catalog No. p-7136192
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T02895G 5 g
T028925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T02895G Supplier TCI America Supplier No. T02895G
Only null left

Chemical Identifiers

CAS 108-40-7
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004843
InChI Key WRXOZRLZDJAYDR-UHFFFAOYSA-N
Synonym m-thiocresol, m-toluenethiol, 3-methylthiophenol, 3-thiocresol, benzenethiol, 3-methyl, m-methylbenzenethiol, m-methylthiophenol, 3-mercaptotoluene, m-tolylmercaptan, m-mercaptotoluene
PubChem CID 7930
IUPAC Name 3-methylbenzene-1-thiol
SMILES CC1=CC=CC(S)=C1

Specifications

Color Yellow
Boiling Point 195°C
Quantity 5 g
UN Number 2810
Formula Weight 124.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material m-Toluenethiol
EINECSNumber (3)-2895
RTECSNumber XT8700000
TSCA Yes