missing translation for 'onlineSavingsMsg'
Learn More

m-Toluamide, 99%

Catalog No. AAB2444222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2444222 100 g
AAB2444214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2444222 Supplier Thermo Scientific Chemicals Supplier No. B2444222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 618-47-3 | C8H9NO | 135.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-47-3
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00007990
InChI Key WGRPQCFFBRDZFV-UHFFFAOYSA-N
Synonym m-toluamide, benzamide, 3-methyl, m-methylbenzamide, 3-methyl-benzamide, benzamide, 3-methyl-9ci, meta-toluamide, 3-methylbenzamide, acmc-1b8qe, 3-methylbenzenecarboximidic acid
PubChem CID 69253
IUPAC Name 3-methylbenzamide
SMILES CC1=CC=CC(=C1)C(=O)N

Specifications

Melting Point 93°C to 96°C
Quantity 100 g
Beilstein 1857374
Formula Weight 135.17
Percent Purity 99%
Chemical Name or Material m-Toluamide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 210-553-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only