accessibility menu, dialog, popup

UserName

CAS RN 618-47-3

IUPAC Name: 3-methylbenzamide
Synonyms: 3-methylbenzamide
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: WGRPQCFFBRDZFV-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results