missing translation for 'onlineSavingsMsg'
Learn More

m-Tolualdehyde (stabilized with HQ) 97.0+%, TCI America™

Catalog No. T02585ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T02585ML 5 mL
T025825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T02585ML Supplier TCI America Supplier No. T02585ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 620-23-5
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00003374
InChI Key OVWYEQOVUDKZNU-UHFFFAOYSA-N
Synonym m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde
PubChem CID 12105
ChEBI CHEBI:28476
IUPAC Name 3-methylbenzaldehyde
SMILES CC1=CC=CC(=C1)C=O

Specifications

Color Yellow
Boiling Point 200°C
Quantity 5 mL
Formula Weight 120.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material m-Tolualdehyde (stabilized with HQ)
EINECSNumber (3)-2659
RTECSNumber CU7033700
TSCA Yes