accessibility menu, dialog, popup

UserName

CH3O

CAS RN 620-23-5

IUPAC Name: 3-methylbenzaldehyde
Synonyms: 3-methylbenzaldehyde M-tolualdehyde
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: OVWYEQOVUDKZNU-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C=O)=C1
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)