missing translation for 'onlineSavingsMsg'
Learn More

m-Tolualdehyde, 98%, stabilized

Catalog No. p-200978
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138990250 25 g Glass bottle
AC138991000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC138990250 Supplier Thermo Scientific Chemicals Supplier No. 138990250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 620-23-5 | C8H8O | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 620-23-5 , 123-31-9
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00003374
InChI Key OVWYEQOVUDKZNU-UHFFFAOYSA-N
Synonym m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde
PubChem CID 12105
ChEBI CHEBI:28476
IUPAC Name 3-methylbenzaldehyde
SMILES CC1=CC=CC(=C1)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown to Yellow
Density 1.0190g/mL
Boiling Point 199°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5400 to 1.542
Linear Formula CH3C6H4CHO
Quantity 25 g
Beilstein 07, 296
Specific Gravity 1.019
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol and ether
Stabilizer 0.1% hydroquinone
Formula Weight 120.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material m-Tolualdehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
May produce an allergic reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
EINECSNumber 210-632-0
TSCA TSCA
Recommended Storage Product darkens on storage

RUO – Research Use Only