Learn More
m-Tolualdehyde, 97%, stab. with 0.1% hydroquinone
CAS: 620-23-5 | C8H8O | 120.151 g/mol
Supplier: Thermo Scientific Chemicals A1531314
Description
m-Tolualdehyde is used as a fragrance agent in food and cosmetics. It is also used as a reagent in the preparation of a wide range of pharmaceutical compounds such as anti-inflammatory agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsm-Tolualdehyde is used as a fragrance agent in food and cosmetics. It is also used as a reagent in the preparation of a wide range of pharmaceutical compounds such as anti-inflammatory agents.
Solubility
Not miscible in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications
| m-Tolualdehyde | |
| 620-23-5 | |
| 198°C to 200°C | |
| Almond-like | |
| 1.541 | |
| 25 g | |
| Air Sensitive | |
| Not miscible in water. | |
| CC1=CC=CC(=C1)C=O | |
| 120.151 | |
| CHEBI:28476 | |
| 97% |
| stabilized with 0.1% hydroquinone | |
| 1.02 | |
| 78°C (172°F) | |
| C8H8O | |
| MFCD00003374 | |
| 741964 | |
| m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde | |
| OVWYEQOVUDKZNU-UHFFFAOYSA-N | |
| 3-methylbenzaldehyde | |
| 12105 | |
| 120.15 |
Chemical Identifiers
| 620-23-5 | |
| 120.151 | |
| OVWYEQOVUDKZNU-UHFFFAOYSA-N | |
| 12105 | |
| 3-methylbenzaldehyde |
| C8H8O | |
| MFCD00003374 | |
| m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde | |
| CHEBI:28476 | |
| CC1=CC=CC(=C1)C=O |
Safety and Handling
GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
EINECSNumber : 210-632-0
RTECSNumber : CU7033700
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only