missing translation for 'onlineSavingsMsg'
Learn More

m-Tolualdehyde, 97%, stab. with 0.1% hydroquinone

Catalog No. AAA1531306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1531306 5 g
AAA1531314 25 g
AAA1531322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1531306 Supplier Thermo Scientific Chemicals Supplier No. A1531306

CAS: 620-23-5 | C8H8O | 120.151 g/mol

m-Tolualdehyde is used as a fragrance agent in food and cosmetics. It is also used as a reagent in the preparation of a wide range of pharmaceutical compounds such as anti-inflammatory agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Tolualdehyde is used as a fragrance agent in food and cosmetics. It is also used as a reagent in the preparation of a wide range of pharmaceutical compounds such as anti-inflammatory agents.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 620-23-5
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00003374
InChI Key OVWYEQOVUDKZNU-UHFFFAOYSA-N
Synonym m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde
PubChem CID 12105
ChEBI CHEBI:28476
IUPAC Name 3-methylbenzaldehyde
SMILES CC1=CC=CC(=C1)C=O

Specifications

Density 1.02
Boiling Point 198°C to 200°C
Flash Point 78°C (172°F)
Odor Almond-like
Refractive Index 1.541
Quantity 5 g
Beilstein 741964
Sensitivity Air Sensitive
Solubility Information Not miscible in water.
Formula Weight 120.15
Percent Purity 97%
Chemical Name or Material m-Tolualdehyde, stabilized with 0.1% hydroquinone
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-632-0
RTECSNumber CU7033700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only