missing translation for 'onlineSavingsMsg'
Learn More

(m-Terphenyl-5'-yl)trimethylsilane 98.0+%, TCI America™

Catalog No. T18711G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18711G Supplier TCI America Supplier No. T18711G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128388-53-4
Molecular Formula C21H22Si
Molecular Weight (g/mol) 302.492
MDL Number MFCD03844807
InChI Key GCYRLUSIWVFXEZ-UHFFFAOYSA-N
Synonym 3,5-Diphenyl-1-trimethylsilylbenzene, Trimethyl(m-terphenyl-5′C-yl)silane
PubChem CID 625570
IUPAC Name (3,5-diphenylphenyl)-trimethylsilane
SMILES C[Si](C)(C)C1=CC(=CC(=C1)C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 81°C
Color White
Quantity 1 g
Formula Weight 302.49
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (m-Terphenyl-5′-yl)trimethylsilane