Learn More
m-Phenylenediamine, 98%
CAS: 108-45-2 | C6H8N2 | 108.144 g/mol
Supplier: Thermo Scientific Chemicals A153390B
Description
m-Phenylenediamine is used in the foaming-type hair dye composition. It is used as a synthetic electronic grade polyimide material and epoxy resin curing agent. It is used also block polymers, textile fibers, urethanes, petroleum additives, rubber chemicals, in corrosion inhibitors, in photography, as reagent for gold & bromine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsm-Phenylenediamine is used in the foaming-type hair dye composition. It is used as a synthetic electronic grade polyimide material and epoxy resin curing agent. It is used also block polymers, textile fibers, urethanes, petroleum additives, rubber chemicals, in corrosion inhibitors, in photography, as reagent for gold & bromine.
Solubility
Soluble in water, methanol, acetone, DMF, MEK, dioxane
Notes
Air & Light Sensitive. Store in the dark. Store under dry argon. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents.
Specifications
| m-Phenylenediamine | |
| 62°C to 66°C | |
| 284°C to 287°C | |
| C6H8N2 | |
| 1000 g | |
| 471357 | |
| 14,7283 | |
| Soluble in water, methanol, acetone, DMF, MEK, dioxane | |
| C1=CC(=CC(=C1)N)N | |
| 108.144 | |
| CHEBI:8092 | |
| 98% |
| 108-45-2 | |
| 1.139 | |
| 187°C (368°F) | |
| MFCD00007799 | |
| UN1673 | |
| Air and light sensitive | |
| m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamine | |
| WZCQRUWWHSTZEM-UHFFFAOYSA-N | |
| benzene-1,3-diamine | |
| 7935 | |
| 108.14 |
Chemical Identifiers
| 108-45-2 | |
| 108.144 | |
| WZCQRUWWHSTZEM-UHFFFAOYSA-N | |
| 7935 | |
| benzene-1,3-diamine |
| C6H8N2 | |
| MFCD00007799 | |
| m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamine | |
| CHEBI:8092 | |
| C1=CC(=CC(=C1)N)N |
Safety and Handling
GHS H Statement
H301-H311-H331-H341-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H319-H341-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLENEDIAMINES
EINECSNumber : 203-584-7
RTECSNumber : SS7700000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only