missing translation for 'onlineSavingsMsg'
Learn More

m-Phenylenediamine, 98%

Catalog No. p-7022519
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1533930 250 g
AAA153390B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1533930 Supplier Thermo Scientific Chemicals Supplier No. A1533930
Only null left

CAS: 108-45-2 | C6H8N2 | 108.144 g/mol

m-Phenylenediamine is used in the foaming-type hair dye composition. It is used as a synthetic electronic grade polyimide material and epoxy resin curing agent. It is used also block polymers, textile fibers, urethanes, petroleum additives, rubber chemicals, in corrosion inhibitors, in photography, as reagent for gold & bromine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Phenylenediamine is used in the foaming-type hair dye composition. It is used as a synthetic electronic grade polyimide material and epoxy resin curing agent. It is used also block polymers, textile fibers, urethanes, petroleum additives, rubber chemicals, in corrosion inhibitors, in photography, as reagent for gold & bromine.

Solubility
Soluble in water, methanol, acetone, DMF, MEK, dioxane

Notes
Air & Light Sensitive. Store in the dark. Store under dry argon. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents.

Chemical Identifiers

CAS 108-45-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00007799
InChI Key WZCQRUWWHSTZEM-UHFFFAOYSA-N
Synonym m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamine
PubChem CID 7935
ChEBI CHEBI:8092
IUPAC Name benzene-1,3-diamine
SMILES C1=CC(=CC(=C1)N)N

Specifications

Melting Point 62°C to 66°C
Density 1.139
Boiling Point 284°C to 287°C
Flash Point 187°C (368°F)
Quantity 250 g
UN Number UN1673
Beilstein 471357
Sensitivity Air and light sensitive
Merck Index 14,7283
Solubility Information Soluble in water, methanol, acetone, DMF, MEK, dioxane
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material m-Phenylenediamine
Show More Show Less
Hazard Category H301+H311+H331-H317-H319-H341-H500
Hazard Statement GHS H Statement
H301-H311-H331-H341-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLENEDIAMINES
EINECSNumber 203-584-7
RTECSNumber SS7700000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only