accessibility menu, dialog, popup

UserName

NH2H2N

CAS RN 108-45-2

IUPAC Name: benzene-1,3-diamine
Synonyms: developer C C.I. developer 11 direct brown GG direct brown BR CI developer 11 3-phenylenediamine developer 11 metaphenylenediamine developer M developer H
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: WZCQRUWWHSTZEM-UHFFFAOYSA-N
SMILES: NC1=CC(N)=CC=C1
Boiling Point 
  • (1)
  • (5)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (6)
  • (6)
Percent Purity 
  • (4)
  • (2)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (4)
  • (7)
  • (1)
  • (5)
  • (2)
  • (4)
  • (6)
  • (6)
  • (4)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (4)
  • (5)