missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Anisic Acid 98.0+%, TCI America™
Supplier: TCI America A1070250G
Specifications
| m-Anisic Acid | |
| 107°C | |
| 172°C | |
| MFCD00002499 | |
| m-anisic acid, m-methoxybenzoic acid, benzoic acid, 3-methoxy, 3-anisic acid, meta-anisic acid, 3-methoxy-benzoic acid, unii-e2i36fh6qz, m-methylsalicylic acid, e2i36fh6qz, chembl22425 | |
| COC1=CC=CC(=C1)C(=O)O | |
| 152.149 | |
| 152.15 | |
| Crystalline Powder |
| 586-38-9 | |
| White-Yellow | |
| C8H8O3 | |
| 250 g | |
| XHQZJYCNDZAGLW-UHFFFAOYSA-N | |
| 3-methoxybenzoic acid | |
| 11461 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 586-38-9 | |
| 152.149 | |
| XHQZJYCNDZAGLW-UHFFFAOYSA-N | |
| 11461 | |
| COC1=CC=CC(=C1)C(=O)O |
| C8H8O3 | |
| MFCD00002499 | |
| m-anisic acid, m-methoxybenzoic acid, benzoic acid, 3-methoxy, 3-anisic acid, meta-anisic acid, 3-methoxy-benzoic acid, unii-e2i36fh6qz, m-methylsalicylic acid, e2i36fh6qz, chembl22425 | |
| 3-methoxybenzoic acid |
Safety and Handling
EINECSNumber : (3)-1659
RTECSNumber : BZ4375000
TSCA : Yes