missing translation for 'onlineSavingsMsg'
Learn More

m-Anisic Acid 98.0+%, TCI America™

Catalog No. p-7117224
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
A107025G 25 g
A1070250G 250 g
2 options

Catalog No. A107025G

Supplier: TCI America A107025G

Only null left

Chemical Identifiers

CAS 586-38-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002499
InChI Key XHQZJYCNDZAGLW-UHFFFAOYSA-N
Synonym m-anisic acid, m-methoxybenzoic acid, benzoic acid, 3-methoxy, 3-anisic acid, meta-anisic acid, 3-methoxy-benzoic acid, unii-e2i36fh6qz, m-methylsalicylic acid, e2i36fh6qz, chembl22425
PubChem CID 11461
IUPAC Name 3-methoxybenzoic acid
SMILES COC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 172°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material m-Anisic Acid
EINECSNumber (3)-1659
RTECSNumber BZ4375000
TSCA Yes