missing translation for 'onlineSavingsMsg'
Learn More

m-(4-Fluorophenoxy)toluene 97.0+%, TCI America™

Catalog No. F03745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03745G Supplier TCI America Supplier No. F03745G
Only null left

Chemical Identifiers

CAS 1514-26-7
Molecular Formula C13H11FO
Molecular Weight (g/mol) 202.228
MDL Number MFCD01321168
InChI Key APVQRVSBMIDSFS-UHFFFAOYSA-N
Synonym 4-Fluoro-3′C-methyldiphenyl Ether, 4-Fluorophenyl m-Tolyl Ether
PubChem CID 20068801
IUPAC Name 1-fluoro-4-(3-methylphenoxy)benzene
SMILES CC1=CC(=CC=C1)OC2=CC=C(C=C2)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 202.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material m-(4-Fluorophenoxy)toluene